Learn More
Cayman Chemical Reuterin, Purity: ≥95%, CAS: 2134-29-4, 100 mg
SDP
Catalog No. 502911753
Encompass_Preferred
A bacterial glycerol metabolite with antibacterial activity; is produced by L. reuteri during exposure to live or UV-killed E. coli; active against E. coli and several species of Bacteroides, Bifidobacterium, and Eubacterium (MICs = 1.9-15 mM) as well as C. difficile and B. vulgatus (MICs = C. difficile at 2.5 mM; alters the bacterial composition of, and VOCs produced by, the mouse fecal microbiome
Catalog No. 50-291-1753 Supplier Cayman Chemical Supplier No. 36300100 MG
A bacterial glycerol metabolite with antibacterial activity; is produced by L. reuteri during exposure to live or UV-killed E. coli; active against E. coli and several species of Bacteroides, Bifidobacterium, and Eubacterium (MICs = 1.9-15 mM) as well as C. difficile and B. vulgatus (MICs = C. difficile at 2.5 mM; alters the bacterial composition of, and VOCs produced by, the mouse fecal microbiome