Learn More
Cayman Chemical RG3039, Purity: ≥98%, CAS: 1005504-62-0, 25 mg
SDP
Catalog No. 502909491
Encompass_Preferred
A DcpS inhibitor (IC50 = 0.069 nM); prevents denervation of the NMJ and decreases in NMJ synaptic transmission and myofiber size of the longissimus muscle in a mouse model of Smn1 knockout-induced SMA at 10 mg/kg; improves survival and motor function in the same model
Catalog No. 50-290-9491 Supplier Cayman Chemical Supplier No. 3658925 MG
A DcpS inhibitor (IC50 = 0.069 nM); prevents denervation of the NMJ and decreases in NMJ synaptic transmission and myofiber size of the longissimus muscle in a mouse model of Smn1 knockout-induced SMA at 10 mg/kg; improves survival and motor function in the same model