Learn More
Cayman Chemical Ribitol, Purity: ≥98%, CAS: 488-81-3, 25 g
SDP
Catalog No. 502937357
Encompass_Preferred
A pentose sugar alcohol; a component of the glycopolymer teichoic acid in bacterial cell walls; levels are increased in the brain, cerebrospinal fluid, urine, and plasma of a patient with RPI deficiency
Catalog No. 50-293-7357 Supplier Cayman Chemical Supplier No. 2736225 G
A pentose sugar alcohol; a component of the glycopolymer teichoic acid in bacterial cell walls; levels are increased in the brain, cerebrospinal fluid, urine, and plasma of a patient with RPI deficiency