Learn More
Cayman Chemical Ridaifen-B, Purity: ≥98%, CAS: 886465-70-9, 5 mg
SDP
Catalog No. 502987227
Encompass_Preferred
An antagonist of ERα (IC50 = 52.4 nM), an inverse agonist of CB2 (Ki = 43.7 nM), and a derivative of tamoxifen; selective for CB2 over CB1 receptors (Ki = 732 nM); designed to be cytotoxic to cancer cells independent of ER binding; it inhibits growth of ER-positive and ER-negative cells in a panel of 39 cancer cell lines (GI50s = 0.20-2.14 µM); induces apoptosis and autophagy in ER-negative Jurkat cells at 0.4 µM; decreases NO production and protein levels of IL-1α and IL-6 in LPS-stimulated RAW 264.7 cells at 1 µM
Catalog No. 50-298-7227 Supplier Cayman Chemical Supplier No. 219665 MG
An antagonist of ERα (IC50 = 52.4 nM), an inverse agonist of CB2 (Ki = 43.7 nM), and a derivative of tamoxifen; selective for CB2 over CB1 receptors (Ki = 732 nM); designed to be cytotoxic to cancer cells independent of ER binding; it inhibits growth of ER-positive and ER-negative cells in a panel of 39 cancer cell lines (GI50s = 0.20-2.14 µM); induces apoptosis and autophagy in ER-negative Jurkat cells at 0.4 µM; decreases NO production and protein levels of IL-1α and IL-6 in LPS-stimulated RAW 264.7 cells at 1 µM