Learn More
Cayman Chemical RMC-4550, Purity: ≥98%, CAS: 2172651-73-7, 5 mg
SDP
Catalog No. 502989794
Encompass_Preferred
A SHP-2 inhibitor (IC50 = 1.55 nM); selective for SHP-2 over a panel of 14 additional protein phosphatases and a panel of 468 protein kinases at 10 µM; inhibits tumor ERK phosphorylation in a KYSE-520 human esophageal cancer xenograft model in a dose-dependent manner
Catalog No. 50-298-9794 Supplier Cayman Chemical Supplier No. 310115 MG
A SHP-2 inhibitor (IC50 = 1.55 nM); selective for SHP-2 over a panel of 14 additional protein phosphatases and a panel of 468 protein kinases at 10 µM; inhibits tumor ERK phosphorylation in a KYSE-520 human esophageal cancer xenograft model in a dose-dependent manner