Learn More
Cayman Chemical RN-18, Purity: ≥98%, CAS: 431980-38-0, 1 mg
SDP
Catalog No. 502993868
Encompass_Preferred
An HIV-1 Vif inhibitor; inhibits Vif-mediated degradation of the DNA editing enzyme APOBEC3G (IC50s = 10-30 µM); inhibits HIV-1 viral replication (IC50s = 4.5 and 10 µM for nonpermissive CEM and H9 cells, respectively, and >100 µM for permissive cells)
Catalog No. 50-299-3868 Supplier Cayman Chemical Supplier No. 233311 MG
An HIV-1 Vif inhibitor; inhibits Vif-mediated degradation of the DNA editing enzyme APOBEC3G (IC50s = 10-30 µM); inhibits HIV-1 viral replication (IC50s = 4.5 and 10 µM for nonpermissive CEM and H9 cells, respectively, and >100 µM for permissive cells)