Learn More
Cayman Chemical RN-486, Purity: ≥98%, CAS: 1242156-23-5, 10 mg
SDP
Catalog No. 502981036
Encompass_Preferred
A selective BTK inhibitor (Kds = 0.31 nM; IC50 = 4 nM); produces anti-inflammatory and bone-protective effects in mouse collagen-induced arthritis and rat adjuvant-induced arthritis models
Catalog No. 50-298-1036 Supplier Cayman Chemical Supplier No. 2053610 MG
A selective BTK inhibitor (Kds = 0.31 nM; IC50 = 4 nM); produces anti-inflammatory and bone-protective effects in mouse collagen-induced arthritis and rat adjuvant-induced arthritis models