Learn More
Cayman Chemical Ro 4929097, Purity: ≥98%, CAS: 847925-91-1, 1 mg
SDP
Catalog No. 502982973
Encompass_Preferred
A potent, selective inhibitor of γ-secretase (IC50 = 4 nM) that blocks both Aβ40 and Notch processing (IC50s = 14 and 5 nM, respectively); orally active against cancer xenograft models, altering gene expression related to angiogenesis and cell differentiation
Catalog No. 50-298-2973 Supplier Cayman Chemical Supplier No. 199961 MG
A potent, selective inhibitor of γ-secretase (IC50 = 4 nM) that blocks both Aβ40 and Notch processing (IC50s = 14 and 5 nM, respectively); orally active against cancer xenograft models, altering gene expression related to angiogenesis and cell differentiation