Learn More
Cayman Chemical ANTIBODY RotlerIn 10mg
SDP
Catalog No. 502955954
Encompass_Preferred
Inhibits PKCδ (IC50 = 3 μM), CAM kinase III, and a wide range of protein kinases, including PRAK and MAPKAP-K2 (IC50s = 1.9 and 5 μM, respectively); stimulates autophagy by targeting a signaling cascade upstream of mTORC1
Catalog No. 50-295-5954 Supplier Cayman Chemical Supplier No. 1200610 MG
Inhibits PKCδ (IC50 = 3 μM), CAM kinase III, and a wide range of protein kinases, including PRAK and MAPKAP-K2 (IC50s = 1.9 and 5 μM, respectively); stimulates autophagy by targeting a signaling cascade upstream of mTORC1