Learn More
Cayman Chemical RR6, Purity: ≥98%, CAS: 1351758-37-6, 5 mg
SDP
Catalog No. 502894026
Encompass_Preferred
A vanin-1 inhibitor (IC50 = 0.54 µM for the recombinant human enzyme); selective for vanin-1 over biotinidase, cathepsin B, cathepsin L, and papain (IC50s = >200 µM for all); active against P. falciparum (IC50 = 14.5 µM); reduces plasma levels of cholesterol and increases plasma levels of free fatty acids in fasted rats at 3 mg/ml in the drinking water; decreases neointima formation and macrophage infiltration in aortic allografts in a rat model of transplant vasculopathy
Catalog No. 50-289-4026 Supplier Cayman Chemical Supplier No. 90014115 MG
A vanin-1 inhibitor (IC50 = 0.54 µM for the recombinant human enzyme); selective for vanin-1 over biotinidase, cathepsin B, cathepsin L, and papain (IC50s = >200 µM for all); active against P. falciparum (IC50 = 14.5 µM); reduces plasma levels of cholesterol and increases plasma levels of free fatty acids in fasted rats at 3 mg/ml in the drinking water; decreases neointima formation and macrophage infiltration in aortic allografts in a rat model of transplant vasculopathy