Learn More
Cayman Chemical ANTIBODY RRx-001 10mg
SDP
Catalog No. 502940051
Encompass_Preferred
An anticancer agent; inhibits growth in a panel of 12 cancer cell lines (IC50s = 1.8-6 μM), an effect that is enhanced in SCC VII cells under hypoxic conditions; increases intracellular ROS levels in HT-29 and SCC VII cells; induces apoptosis in SCC VII, HL-60, and Daudi cells at 5, 5, and 4 μM, respectively; reduces tumor growth and enhances radiation-induced tumor growth reduction in an SCC VII murine squamous cell carcinoma model at 5 mg/kg per day for five days; pre-incubation of red blood cells and hemolysate at 3 mM increases nitrite-induced NO release under hypoxic conditions; decreases parasitemia and increases survival in a mouse model of P. berghei ANKA infection at 10 mg/kg
Catalog No. 50-294-0051 Supplier Cayman Chemical Supplier No. 2884610 MG
An anticancer agent; inhibits growth in a panel of 12 cancer cell lines (IC50s = 1.8-6 μM), an effect that is enhanced in SCC VII cells under hypoxic conditions; increases intracellular ROS levels in HT-29 and SCC VII cells; induces apoptosis in SCC VII, HL-60, and Daudi cells at 5, 5, and 4 μM, respectively; reduces tumor growth and enhances radiation-induced tumor growth reduction in an SCC VII murine squamous cell carcinoma model at 5 mg/kg per day for five days; pre-incubation of red blood cells and hemolysate at 3 mM increases nitrite-induced NO release under hypoxic conditions; decreases parasitemia and increases survival in a mouse model of P. berghei ANKA infection at 10 mg/kg