Learn More
Cayman Chemical ANTIBODY RSV-604 25mg
SDP
Catalog No. 502941030
Encompass_Preferred
An inhibitor of RSV; binds to RSV nucleoprotein (Kd = 1.31 μM); reduces plaque formation by RSS, Long, A2, and B RSV strains (EC50s = 0.9, 0.8, 0.5, and 0.6 μM, respectively); active against 40 RSV clinical isolates (mean EC50 = 0.8 μM); increases viability of RSV-infected HEp-2 cells (EC50 = 0.86 μM); inhibits RSV replication in HAE cell monolayers following mucosal inoculation
Catalog No. 50-294-1030 Supplier Cayman Chemical Supplier No. 2904125 MG
An inhibitor of RSV; binds to RSV nucleoprotein (Kd = 1.31 μM); reduces plaque formation by RSS, Long, A2, and B RSV strains (EC50s = 0.9, 0.8, 0.5, and 0.6 μM, respectively); active against 40 RSV clinical isolates (mean EC50 = 0.8 μM); increases viability of RSV-infected HEp-2 cells (EC50 = 0.86 μM); inhibits RSV replication in HAE cell monolayers following mucosal inoculation