Learn More
Cayman Chemical RU-521, Purity: ≥98%, CAS: 2262452-06-0, 1 mg
SDP
Catalog No. 502991714
Encompass_Preferred
A cGAS inhibitor (IC50 = 0.11 µM); inhibits dsDNA-induced IFN-β1-dependent gene expression in a reporter assay; reduces basal Ifnb1 mRNA expression in BMDMs generated from the Trex1-/- mouse model of Aicardi-Goutières syndrome
Catalog No. 50-299-1714 Supplier Cayman Chemical Supplier No. 317651 MG
A cGAS inhibitor (IC50 = 0.11 µM); inhibits dsDNA-induced IFN-β1-dependent gene expression in a reporter assay; reduces basal Ifnb1 mRNA expression in BMDMs generated from the Trex1-/- mouse model of Aicardi-Goutières syndrome