Learn More
Cayman Chemical ANTIBODY RY785 500ug
SDP
Catalog No. 502982785
Encompass_Preferred
A selective inhibitor of Kv2.1 voltage-gated potassium channels (IC50 = 0.05 µM)
Catalog No. 50-298-2785 Supplier Cayman Chemical Supplier No. 19813500 UG
A selective inhibitor of Kv2.1 voltage-gated potassium channels (IC50 = 0.05 µM)