Learn More
Cayman Chemical S 2101, Purity: ≥98%, CAS: 1239262-36-2, 10 mg
SDP
Catalog No. 502981353
Encompass_Preferred
An LSD1 inhibitor (Ki = 0.61 µM); selective for LSD1 over MAO-A and MAO-B (Kis = 17 and 110 µM, respectively)
Catalog No. 50-298-1353 Supplier Cayman Chemical Supplier No. 1980210 MG
An LSD1 inhibitor (Ki = 0.61 µM); selective for LSD1 over MAO-A and MAO-B (Kis = 17 and 110 µM, respectively)