Learn More
Cayman Chemical ANTIBODY S1QEL1 1 500ug
SDP
Catalog No. 502982742
Encompass_Preferred
A suppressor of IQ electron leak (IC50 = 70 nM); diminishes endogenous oxidative damage in primary astrocytes, reduces stress-induced signaling in a cardiomyocyte cell model, and protects against ischemia-reperfusion injury in perfused mouse heart
Catalog No. 50-298-2742 Supplier Cayman Chemical Supplier No. 20982500 UG
A suppressor of IQ electron leak (IC50 = 70 nM); diminishes endogenous oxidative damage in primary astrocytes, reduces stress-induced signaling in a cardiomyocyte cell model, and protects against ischemia-reperfusion injury in perfused mouse heart