Learn More
Cayman Chemical Salbutamol, Purity: ≥98%, CAS: 18559-94-9, 50 mg
SDP
Catalog No. 502982497
Encompass_Preferred
A β2-AR agonist; induces bronchodilation in isolated guinea pig trachea rings (EC50 = 19 nM); selectively binds to the β2-AR over the β1-AR and β3-AR in CHO cells expressing the human receptors (Kds = 0.76, 21, and 46 μM, respectively); reduces acetylcholine-induced bronchospasm in anesthetized guinea pigs at 25 and 50 μg/kg, i.v.; reduces the response of bronchial muscle to efferent vagal stimulation in anesthetized cats and dogs; reduces transpulmonary pressure in recurrent airway obstruction-affected horses (ED50 = 43.6 μg/animal) when administered via nebulization
Catalog No. 50-298-2497 Supplier Cayman Chemical Supplier No. 2100350 MG
A β2-AR agonist; induces bronchodilation in isolated guinea pig trachea rings (EC50 = 19 nM); selectively binds to the β2-AR over the β1-AR and β3-AR in CHO cells expressing the human receptors (Kds = 0.76, 21, and 46 μM, respectively); reduces acetylcholine-induced bronchospasm in anesthetized guinea pigs at 25 and 50 μg/kg, i.v.; reduces the response of bronchial muscle to efferent vagal stimulation in anesthetized cats and dogs; reduces transpulmonary pressure in recurrent airway obstruction-affected horses (ED50 = 43.6 μg/animal) when administered via nebulization