Learn More
Cayman Chemical SAR131675, Purity: ≥98%, CAS: 1433953-83-3, 1 mg
SDP
Catalog No. 502939297
Encompass_Preferred
An ATP-competitive inhibitor of VEGFR3 (IC50 = 23 nM); selective for VEGFR3 over VEGFR2 and VEGFR1 (IC50s = 230 and >3,000 nM, respectively); inhibits survival of human lymphatic cells cultured with the VEGFR3 ligands VEGF-C and VEGF-D over the non-specific ligand VEGF-A in vitro (IC50s = 14, 17, and 664 nM, respectively); decreases VEGF-C and VEGF-A-induced migration of HMVECs at 100 and 300 nM; reduces tumor size, decreases the number of pancreatic angiogenic islets, and increases survival in the RIP1-Tag2 transgenic mouse model of pancreatic neuroendocrine tumors at 100 mg/kg; decreases tumor size, levels of VEGFR3 in mammary tumor lysates, number of tumor-associated macrophages in mammary tumors, and the number of lung metastases in a 4T1 mouse allograft model at 100 mg/kg
Catalog No. 50-293-9297 Supplier Cayman Chemical Supplier No. 283851 MG
An ATP-competitive inhibitor of VEGFR3 (IC50 = 23 nM); selective for VEGFR3 over VEGFR2 and VEGFR1 (IC50s = 230 and >3,000 nM, respectively); inhibits survival of human lymphatic cells cultured with the VEGFR3 ligands VEGF-C and VEGF-D over the non-specific ligand VEGF-A in vitro (IC50s = 14, 17, and 664 nM, respectively); decreases VEGF-C and VEGF-A-induced migration of HMVECs at 100 and 300 nM; reduces tumor size, decreases the number of pancreatic angiogenic islets, and increases survival in the RIP1-Tag2 transgenic mouse model of pancreatic neuroendocrine tumors at 100 mg/kg; decreases tumor size, levels of VEGFR3 in mammary tumor lysates, number of tumor-associated macrophages in mammary tumors, and the number of lung metastases in a 4T1 mouse allograft model at 100 mg/kg