Learn More
Cayman Chemical Sarcosine, Purity: ≥95%, CAS: 107-97-1, 100 g
SDP
Catalog No. 502936766
Encompass_Preferred
An intermediate in the biosynthesis of glycine; a selective inhibitor of GlyT1 (IC50s = 91 and >1,000 µM for GlyT1 and GlyT2, respectively); increased in prostate cancer cell lines with an invasive phenotype compared with non-invasive cell lines; increases in a progressive fashion in prostate cancer with 0, 42, and 79% of benign, localized, and metastatic prostate tissue samples containing increased levels of sarcosine
Catalog No. 50-293-6766 Supplier Cayman Chemical Supplier No. 26799100 G
An intermediate in the biosynthesis of glycine; a selective inhibitor of GlyT1 (IC50s = 91 and >1,000 µM for GlyT1 and GlyT2, respectively); increased in prostate cancer cell lines with an invasive phenotype compared with non-invasive cell lines; increases in a progressive fashion in prostate cancer with 0, 42, and 79% of benign, localized, and metastatic prostate tissue samples containing increased levels of sarcosine