Learn More
Cayman Chemical SB-332235, Purity: ≥95%, CAS: 276702-15-9, 1 mg
SDP
Catalog No. 502950009
Encompass_Preferred
A CXCR2 antagonist (IC50 = 9.3 nM); selective for CXCR2 over CXCR1 (IC50 = 9,633 nM); inhibits CXCL1, -5, or -8-induced migration of isolated human peripheral blood monocytes at 10 ng/ml; decreases cigarette smoke-induced increases in BALF neutrophil infiltration at 1 mg/kg; reduces Aβ42-induced increases in neuronal levels of 4-HNE in a rat model of Alzheimer's disease
Catalog No. 50-295-0009 Supplier Cayman Chemical Supplier No. 328691 MG
A CXCR2 antagonist (IC50 = 9.3 nM); selective for CXCR2 over CXCR1 (IC50 = 9,633 nM); inhibits CXCL1, -5, or -8-induced migration of isolated human peripheral blood monocytes at 10 ng/ml; decreases cigarette smoke-induced increases in BALF neutrophil infiltration at 1 mg/kg; reduces Aβ42-induced increases in neuronal levels of 4-HNE in a rat model of Alzheimer's disease