Learn More
Cayman Chemical SB-4, Purity: ≥98%, CAS: 100874-08-6, 5 mg
SDP
Catalog No. 502916317
Encompass_Preferred
An activator of BMP4 signaling; induces BMP4 signaling in a reporter assay using HEK293 cells expressing a recombinant human BMP4 response element (EC50 = 74 nM); increases phosphorylation of SMAD1, -5, and -9 in HEK293 cells at 10 µM; induces Blimp1 and Tfap2c expression in mouse embryonic stem cells
Catalog No. 50-291-6317 Supplier Cayman Chemical Supplier No. 377485 MG
An activator of BMP4 signaling; induces BMP4 signaling in a reporter assay using HEK293 cells expressing a recombinant human BMP4 response element (EC50 = 74 nM); increases phosphorylation of SMAD1, -5, and -9 in HEK293 cells at 10 µM; induces Blimp1 and Tfap2c expression in mouse embryonic stem cells