Learn More
Cayman Chemical SB-611812, Purity: ≥98%, CAS: 345892-71-9, 1 mg
SDP
Catalog No. 502949911
Encompass_Preferred
An antagonist of UTR; binds to recombinant rat UTR (Ki = 121 nM) and inhibits contraction of isolated rat aorta induced by urotensin II (pA2 = 6.59) but not potassium chloride, endothelin-1, angiotensin II, or phenylephrine; decreases myocardial interstitial fibrosis and the ratio of collagen I to collagen III in a rat model of ischemia induced by ligation of the LAD artery (30 mg/kg)
Catalog No. 50-294-9911 Supplier Cayman Chemical Supplier No. 332471 MG
An antagonist of UTR; binds to recombinant rat UTR (Ki = 121 nM) and inhibits contraction of isolated rat aorta induced by urotensin II (pA2 = 6.59) but not potassium chloride, endothelin-1, angiotensin II, or phenylephrine; decreases myocardial interstitial fibrosis and the ratio of collagen I to collagen III in a rat model of ischemia induced by ligation of the LAD artery (30 mg/kg)