Learn More
Cayman Chemical SB-674042, Purity: ≥98%, CAS: 483313-22-0, 1 mg
SDP
Catalog No. 502985057
Encompass_Preferred
A potent and selective non-peptide OX1R antagonist (Kd = 5.03 and 3.76 nM in whole cell binding and membrane-based assays, respectively); >100-fold selective for OX1 over OX2 in calcium mobilization; no significant affinity for serotonergic, dopaminergic, adrenergic, or purinergic receptors
Catalog No. 50-298-5057 Supplier Cayman Chemical Supplier No. 212701 MG
A potent and selective non-peptide OX1R antagonist (Kd = 5.03 and 3.76 nM in whole cell binding and membrane-based assays, respectively); >100-fold selective for OX1 over OX2 in calcium mobilization; no significant affinity for serotonergic, dopaminergic, adrenergic, or purinergic receptors