Learn More
Cayman Chemical SBP-7455, Purity: ≥98%, CAS: 1884222-74-5, 25 mg
SDP
Catalog No. 502947424
Encompass_Preferred
A dual inhibitor of ULK1 and ULK2 (IC50s = 13 and 476 nM, respectively, for the recombinant human enzymes in cell-free assays); inhibits increases in autophagic flux and induces apoptosis in serum-deprived MDA-MB-468 breast cancer cells at 10 µM; reduces the viability of MDA-MB-468 cells at 0.19 µM alone or in combination with olaparib
Catalog No. 50-294-7424 Supplier Cayman Chemical Supplier No. 3298825 MG
A dual inhibitor of ULK1 and ULK2 (IC50s = 13 and 476 nM, respectively, for the recombinant human enzymes in cell-free assays); inhibits increases in autophagic flux and induces apoptosis in serum-deprived MDA-MB-468 breast cancer cells at 10 µM; reduces the viability of MDA-MB-468 cells at 0.19 µM alone or in combination with olaparib