Learn More
Cayman Chemical SCH 442416, Purity: ≥98%, CAS: 316173-57-6, 25 mg
SDP
Catalog No. 502987667
Encompass_Preferred
A potent, brain-permeable, and selective A2AR antagonist; selective for A2AR over A1R, A2BR, and A3R (Kis = 0.048, 1,111, >10,000, and >10,000 nM, respectively) using human recombinant receptors; selective for A2AR over A1R and A3R in rat cerebral membranes (Kis = 0.50, 1,815, and >10,000 nM, respectively); is taken up preferentially by liver, adrenal gland, kidney, and lungs, and by the striatum in the brain; attenuates adenosine-mediated and A2AR agonist-induced arteriole dilation at a concentration of 10 μM
Catalog No. 50-298-7667 Supplier Cayman Chemical Supplier No. 2223325 MG
A potent, brain-permeable, and selective A2AR antagonist; selective for A2AR over A1R, A2BR, and A3R (Kis = 0.048, 1,111, >10,000, and >10,000 nM, respectively) using human recombinant receptors; selective for A2AR over A1R and A3R in rat cerebral membranes (Kis = 0.50, 1,815, and >10,000 nM, respectively); is taken up preferentially by liver, adrenal gland, kidney, and lungs, and by the striatum in the brain; attenuates adenosine-mediated and A2AR agonist-induced arteriole dilation at a concentration of 10 μM