Learn More
Cayman Chemical Scopoletin, Purity: ≥98%, CAS: 92-61-5, 50 mg
SDP
Catalog No. 502980661
Encompass_Preferred
A natural coumarin that binds iron; inhibits aldose reductase activity in galactose-fed rats, upregulates PPARγ expression, and triggers phosphorylation of Nrf2 in animals
Catalog No. 50-298-0661 Supplier Cayman Chemical Supplier No. 2004250 MG
A natural coumarin that binds iron; inhibits aldose reductase activity in galactose-fed rats, upregulates PPARγ expression, and triggers phosphorylation of Nrf2 in animals