Learn More
Cayman Chemical SEN-12333, Purity: ≥98%, CAS: 874450-44-9, 100 mg
SDP
Catalog No. 502911921
Encompass_Preferred
An agonist of α7 nAChRs (Ki = 0.26 µM); selective for α7 nAChRs over α1-, α3-, and α4β2 subunit-containing nAChRs and the 5-HT3 receptor, as well as a panel of 70 additional neurotransmitter, growth factor, and peptide receptors at 10 µM; activates the α7 nAChR in GH4C1 cells expressing the rat receptor (EC50 = 1.6 µM); topical application prevents decreases in the number of retinal ganglion cells in a rat model of glaucoma; reverses scopolamine- or MK-801-induced memory deficits in the novel objection recognition test in rats at 3 mg/kg
Catalog No. 50-291-1921 Supplier Cayman Chemical Supplier No. 36123100 MG
An agonist of α7 nAChRs (Ki = 0.26 µM); selective for α7 nAChRs over α1-, α3-, and α4β2 subunit-containing nAChRs and the 5-HT3 receptor, as well as a panel of 70 additional neurotransmitter, growth factor, and peptide receptors at 10 µM; activates the α7 nAChR in GH4C1 cells expressing the rat receptor (EC50 = 1.6 µM); topical application prevents decreases in the number of retinal ganglion cells in a rat model of glaucoma; reverses scopolamine- or MK-801-induced memory deficits in the novel objection recognition test in rats at 3 mg/kg