Learn More
Cayman Chemical SET2, Purity: ≥98%, CAS: 2313525-20-9, 50 mg
SDP
Catalog No. 502944472
Encompass_Preferred
A TRPV2 inhibitor (IC50 = 0.46 µM); selectively inhibits 2-APB-induced currents in a patch-clamp assay using HEK293T cells overexpressing TRPV2 at 0.3 or 1 µM over HEK293T cells overexpressing TRPV1 or TRPV3 at 3 or 10 µM; reduces LPA-induced migration in PC-3M prostate cancer cells at 20 µM; reduces LPA-induced increases in current in a patch-clamp assay using HEK293T cells
Catalog No. 50-294-4472 Supplier Cayman Chemical Supplier No. 4036050 MG
A TRPV2 inhibitor (IC50 = 0.46 µM); selectively inhibits 2-APB-induced currents in a patch-clamp assay using HEK293T cells overexpressing TRPV2 at 0.3 or 1 µM over HEK293T cells overexpressing TRPV1 or TRPV3 at 3 or 10 µM; reduces LPA-induced migration in PC-3M prostate cancer cells at 20 µM; reduces LPA-induced increases in current in a patch-clamp assay using HEK293T cells