Learn More
Cayman Chemical ANTIBODY SetosusIn 500ug
SDP
Catalog No. 503015752
Encompass_Preferred
A fungal metabolite; reduces Aβ aggregate-induced neurotoxicity in PC12 cells (EC50 = 112.6 μM); induces tremors in mice at 30 mg/kg
Catalog No. 50-301-5752 Supplier Cayman Chemical Supplier No. 25484500 UG
A fungal metabolite; reduces Aβ aggregate-induced neurotoxicity in PC12 cells (EC50 = 112.6 μM); induces tremors in mice at 30 mg/kg