Learn More
Cayman Chemical SGC3027, Purity: ≥98%, CAS: 2624313-13-7, 1 mg
SDP
Catalog No. 502932661
Encompass_Preferred
A prodrug form of the PRMT7 inhibitor SGC8158; converted by intracellular reductases to SGC8158; inhibits methylation of Hsp70 in vitro (IC50 = 2.4 µM)
Catalog No. 50-293-2661 Supplier Cayman Chemical Supplier No. 253251 MG
A prodrug form of the PRMT7 inhibitor SGC8158; converted by intracellular reductases to SGC8158; inhibits methylation of Hsp70 in vitro (IC50 = 2.4 µM)