Learn More
Cayman Chemical SGI-7079, Purity: ≥98%, CAS: 1239875-86-5, 5 mg
SDP
Catalog No. 502981351
Encompass_Preferred
An Axl kinase inhibitor; inhibits proliferation of inflammatory breast cancer cells (IC50s = 0.43 and 0.15 µM for SUM149 and KPL-4 cells, respectively); synergistically increases the potency of erlotinib on EGFR inhibition; dose-dependently inhibits tumor growth in a mouse xenograft model of non-small cell lung cancer
Catalog No. 50-298-1351 Supplier Cayman Chemical Supplier No. 201115 MG
An Axl kinase inhibitor; inhibits proliferation of inflammatory breast cancer cells (IC50s = 0.43 and 0.15 µM for SUM149 and KPL-4 cells, respectively); synergistically increases the potency of erlotinib on EGFR inhibition; dose-dependently inhibits tumor growth in a mouse xenograft model of non-small cell lung cancer