Learn More
Cayman Chemical SIBA, Purity: ≥98%, CAS: 35899-54-8, 50 mg
SDP
Catalog No. 502915971
Encompass_Preferred
A derivative of SAH; reduces viral infectivity in HEp-2 cells infected with HSV-1 at 1 mM; inhibits the RSV-induced oncogenic transformation of primary chicken embryo fibroblasts at 0.5 and 1 mM; decreases the number of metastases in a model of rhabdomyosarcoma in rats fed a low methionine diet at 15 mg/kg three times weekly
Catalog No. 50-291-5971 Supplier Cayman Chemical Supplier No. 3875850 MG
A derivative of SAH; reduces viral infectivity in HEp-2 cells infected with HSV-1 at 1 mM; inhibits the RSV-induced oncogenic transformation of primary chicken embryo fibroblasts at 0.5 and 1 mM; decreases the number of metastases in a model of rhabdomyosarcoma in rats fed a low methionine diet at 15 mg/kg three times weekly