Learn More
Cayman Chemical Siremadlin, Purity: ≥98%, CAS: 1448867-41-1, 1 mg
SDP
Catalog No. 502916768
Encompass_Preferred
A MDM2-p53 protein-protein interaction inhibitor (IC50 = 0.21 nM); selectively inhibits the MDM2-p53 protein-protein interaction over MDM4-p53, YAP1-TEAD4, PCSK9-LDLR, and Bcl-2-Bak interactions in TR-FRET assays (IC50s = 3.3, >150, >50, and >50 µM, respectively); inhibits the growth of SJSA-1 cells (GI50 = 38 nM); induces apoptosis in SJSA-1 cells at 100 nM; induces tumor regression and prevents tumor regrowth in an SJSA-1 rat xenograft model and an HSAX2655 PDX rat liposarcoma model at 27 mg/kg
Catalog No. 50-291-6768 Supplier Cayman Chemical Supplier No. 385161 MG
A MDM2-p53 protein-protein interaction inhibitor (IC50 = 0.21 nM); selectively inhibits the MDM2-p53 protein-protein interaction over MDM4-p53, YAP1-TEAD4, PCSK9-LDLR, and Bcl-2-Bak interactions in TR-FRET assays (IC50s = 3.3, >150, >50, and >50 µM, respectively); inhibits the growth of SJSA-1 cells (GI50 = 38 nM); induces apoptosis in SJSA-1 cells at 100 nM; induces tumor regression and prevents tumor regrowth in an SJSA-1 rat xenograft model and an HSAX2655 PDX rat liposarcoma model at 27 mg/kg