Learn More
Cayman Chemical Sisomicin, Purity: ≥95%, CAS: 32385-11-8, 100 mg
SDP
Catalog No. 502984075
Encompass_Preferred
A broad-spectrum aminoglycoside antibiotic; active against Gram-positive and Gram-negative bacteria including B. subtilis, S. aureus, E. coli, and P. aeruginosa, among others (MICs = 0.01-0.75 µg/ml); protective against Gram-positive and Gram-negative bacterial infections in mice (PD50s = 0.5-6 mg/kg)
Catalog No. 50-298-4075 Supplier Cayman Chemical Supplier No. 21046100 MG
A broad-spectrum aminoglycoside antibiotic; active against Gram-positive and Gram-negative bacteria including B. subtilis, S. aureus, E. coli, and P. aeruginosa, among others (MICs = 0.01-0.75 µg/ml); protective against Gram-positive and Gram-negative bacterial infections in mice (PD50s = 0.5-6 mg/kg)