Learn More
Cayman Chemical SKPin C1, Purity: ≥98%, CAS: 432001-69-9, 25 mg
SDP
Catalog No. 502935369
Encompass_Preferred
An inhibitor of Skp2-mediated p27 degradation; blocks ubiquitylation of p27 in a Cks1-dependent manner in vitro at 50 μM; increases nuclear accumulation of p27 in ECC-1 endometrial carcinoma cells
Catalog No. 50-293-5369 Supplier Cayman Chemical Supplier No. 2614125 MG
An inhibitor of Skp2-mediated p27 degradation; blocks ubiquitylation of p27 in a Cks1-dependent manner in vitro at 50 μM; increases nuclear accumulation of p27 in ECC-1 endometrial carcinoma cells