Learn More
Cayman Chemical ANTIBODY SMU127 500ug
SDP
Catalog No. 502950281
Encompass_Preferred
An agonist of TLR1/2; induces NF-κB signaling in cells expressing human TLR2 (EC50 = 0.55 µM) but not cells expressing human TLR3, -4, -5, -7, or -8 from 0.1-100 µM; induces the production of TNF-α in isolated human PBMCs from 0.01-1 µM; reduces tumor volume in a 4T1 murine mammary carcinoma model at 0.1 mg/animal
Catalog No. 50-295-0281 Supplier Cayman Chemical Supplier No. 33147500 UG
An agonist of TLR1/2; induces NF-κB signaling in cells expressing human TLR2 (EC50 = 0.55 µM) but not cells expressing human TLR3, -4, -5, -7, or -8 from 0.1-100 µM; induces the production of TNF-α in isolated human PBMCs from 0.01-1 µM; reduces tumor volume in a 4T1 murine mammary carcinoma model at 0.1 mg/animal