Learn More
Cayman Chemical SNAP 94847, Purity: ≥98%, CAS: 487051-12-7, 25 mg
SDP
Catalog No. 502910365
Encompass_Preferred
An antagonist of MCH1 (Ki = 2.2 nM for the recombinant rat receptor); selective for MHC1 over α1A-adrenergic and dopamine D2 receptors (Kis = 180 and 7,400 nM, respectively, for the recombinant human receptors); increases the time mice spend in the light side of the light/dark exploration test at 20 mg/kg; reduces food intake and body weight, as well as plasma levels of glucose, insulin, and triglycerides, in a mouse model of high-fat diet-induced obesity at 5 mg/kg in combination with rimonabant
Catalog No. 50-291-0365 Supplier Cayman Chemical Supplier No. 3510525 MG
An antagonist of MCH1 (Ki = 2.2 nM for the recombinant rat receptor); selective for MHC1 over α1A-adrenergic and dopamine D2 receptors (Kis = 180 and 7,400 nM, respectively, for the recombinant human receptors); increases the time mice spend in the light side of the light/dark exploration test at 20 mg/kg; reduces food intake and body weight, as well as plasma levels of glucose, insulin, and triglycerides, in a mouse model of high-fat diet-induced obesity at 5 mg/kg in combination with rimonabant