Learn More
Cayman Chemical SOBRAC, Purity: ≥95%, CAS: 362678-28-2, 1 mg
SDP
Catalog No. 502896031
Encompass_Preferred
An inhibitor of acid ceramidase (Ki = 29.7 nM)
Catalog No. 50-289-6031 Supplier Cayman Chemical Supplier No. 90039271 MG
An inhibitor of acid ceramidase (Ki = 29.7 nM)