Learn More
Cayman Chemical Sparsentan, Purity: ≥98%, CAS: 254740-64-2, 25 mg
SDP
Catalog No. 502947292
Encompass_Preferred
A dual antagonist of AT1 and ETA (Kis = 0.8 and 9.3 nM, respectively, for the human receptors); selective for these receptors over AT2 and ETB receptors (Kis = >10 µM for both); inhibits angiotensin II-induced pressor responses in conscious normotensive rats (ED50 = 3.6 µmol/kg, p.o); reduces mean arterial pressure in spontaneously hypertensive rats at 10, 30, or 100 µmol/kg per day, p.o.; reduces albuminuria and inhibits development of glomerulosclerosis in the gddY mouse model of IgA nephropathy when administered in the diet at 1,800 ppm
Catalog No. 50-294-7292 Supplier Cayman Chemical Supplier No. 3342925 MG
A dual antagonist of AT1 and ETA (Kis = 0.8 and 9.3 nM, respectively, for the human receptors); selective for these receptors over AT2 and ETB receptors (Kis = >10 µM for both); inhibits angiotensin II-induced pressor responses in conscious normotensive rats (ED50 = 3.6 µmol/kg, p.o); reduces mean arterial pressure in spontaneously hypertensive rats at 10, 30, or 100 µmol/kg per day, p.o.; reduces albuminuria and inhibits development of glomerulosclerosis in the gddY mouse model of IgA nephropathy when administered in the diet at 1,800 ppm