Learn More
Cayman Chemical Spermidine, Purity: ≥98%, CAS: 124-20-9, 25 g
SDP
Catalog No. 502973097
Encompass_Preferred
An endogenous polyamine; inhibits the activity of the human inward-rectifying potassium channel Kir2.3 in a patch-clamp assay at 25 µM; induces autophagy in HeLa cells at 100 µM; increases the lifespan of D. melanogaster, yeast, and C. elegans; reduces demyelination of the optic nerve and disease severity in a mouse model of EAE when administered in the drinking water at 30 mM; reduces increases in blood pressure, left ventricular posterior wall thickness, and heart weight in salt-sensitive Dahl rats fed a high-salt diet
Catalog No. 50-297-3097 Supplier Cayman Chemical Supplier No. 1491825 G
An endogenous polyamine; inhibits the activity of the human inward-rectifying potassium channel Kir2.3 in a patch-clamp assay at 25 µM; induces autophagy in HeLa cells at 100 µM; increases the lifespan of D. melanogaster, yeast, and C. elegans; reduces demyelination of the optic nerve and disease severity in a mouse model of EAE when administered in the drinking water at 30 mM; reduces increases in blood pressure, left ventricular posterior wall thickness, and heart weight in salt-sensitive Dahl rats fed a high-salt diet