Learn More
Cayman Chemical SPI 112, Purity: ≥95%, CAS: 1051387-90-6, 100 mg
SDP
Catalog No. 502988503
Encompass_Preferred
A cell-impermeable inhibitor of SHP-2 in cell-free assays (IC50 = 1 µM); selectively inhibits SHP-2 over SHP-1 and PTP1B (IC50s = 18.3 and 14.5 µM, respectively)
Catalog No. 50-298-8503 Supplier Cayman Chemical Supplier No. 29474100 MG
A cell-impermeable inhibitor of SHP-2 in cell-free assays (IC50 = 1 µM); selectively inhibits SHP-2 over SHP-1 and PTP1B (IC50s = 18.3 and 14.5 µM, respectively)