Learn More
Cayman Chemical ANTIBODY SpInorphIn 500ug
SDP
Catalog No. 502988439
Encompass_Preferred
A peptide inhibitor of enkephalin-degrading enzymes; inhibits aminopeptidase, dipeptidyl aminopeptidase, ACE, and enkephalinase (IC50s = 3.3, 1.4, 2.4, and 10 UG/ml, respectively, for the monkey brain enzymes); selective for these enzymes over human serum aminopeptidase A (IC50 = >100 UG/ml), as well as porcine kidney aminopeptidase B, aminopeptidase M, and DPP-1, -2, -3, and -4 (IC50s = >55 UG/ml for all); inhibits chemotaxis, production of ROS, and exocytosis of glucuronidase and collagenase in PMNs; potentiates enkephalin-induced action potentials in rat hippocampal slices; enhances the antinociceptive effects of Leu-enkephalin in mice at 5 UG/animal
Catalog No. 50-298-8439 Supplier Cayman Chemical Supplier No. 29914500 UG
A peptide inhibitor of enkephalin-degrading enzymes; inhibits aminopeptidase, dipeptidyl aminopeptidase, ACE, and enkephalinase (IC50s = 3.3, 1.4, 2.4, and 10 UG/ml, respectively, for the monkey brain enzymes); selective for these enzymes over human serum aminopeptidase A (IC50 = >100 UG/ml), as well as porcine kidney aminopeptidase B, aminopeptidase M, and DPP-1, -2, -3, and -4 (IC50s = >55 UG/ml for all); inhibits chemotaxis, production of ROS, and exocytosis of glucuronidase and collagenase in PMNs; potentiates enkephalin-induced action potentials in rat hippocampal slices; enhances the antinociceptive effects of Leu-enkephalin in mice at 5 UG/animal