Learn More
Cayman Chemical Spinetoram, Purity: ≥95% (mixture), CAS: 935545-74-7, 1 mg
SDP
Catalog No. 502935147
Encompass_Preferred
An insecticide; induces mortality in H. armigera third, fourth, and fifth instar larvae at 0.19 and 0.36 mg/kg in the diet; induces mortality of wild-type D. melanogaster but not D. melanogaster containing a genetic mutation in the Dα6 subunit of the nAChR (LC50s = 0.025 and 4.4 µg/cm2, respectively, in the diet)
Catalog No. 50-293-5147 Supplier Cayman Chemical Supplier No. 266231 MG
An insecticide; induces mortality in H. armigera third, fourth, and fifth instar larvae at 0.19 and 0.36 mg/kg in the diet; induces mortality of wild-type D. melanogaster but not D. melanogaster containing a genetic mutation in the Dα6 subunit of the nAChR (LC50s = 0.025 and 4.4 µg/cm2, respectively, in the diet)