Learn More
Cayman Chemical ANTIBODY SR 11302 1mg
SDP
Catalog No. 502973311
Encompass_Preferred
A synthetic retinoid that inhibits AP-1 activity without activating transcription through RARE; significantly suppresses both AP-1 activation and phorbol ester-induced papilloma formation in mice when applied topically (0.1 µM)
Catalog No. 50-297-3311 Supplier Cayman Chemical Supplier No. 163381 MG
Add to Cart
Add to Cart
A synthetic retinoid that inhibits AP-1 activity without activating transcription through RARE; significantly suppresses both AP-1 activation and phorbol ester-induced papilloma formation in mice when applied topically (0.1 µM)