Learn More
Cayman Chemical SR 12460, Purity: ≥98%, CAS: 2055101-66-9, 10 mg
SDP
Catalog No. 502990084
Encompass_Preferred
An IKKβ NBD mimetic; inhibits the protein-protein interaction between NEMO/IKKγ and IKKβ; inhibits TNF-α-induced NF-κB activation in a reporter assay from 25-150 µM; reduces LPS-induced IL-6 production in RAW 264.7 cells; improves grip strength in the mdx mouse model of Duchenne muscular dystrophy at 30 mg/kg
Catalog No. 50-299-0084 Supplier Cayman Chemical Supplier No. 3106610 MG
An IKKβ NBD mimetic; inhibits the protein-protein interaction between NEMO/IKKγ and IKKβ; inhibits TNF-α-induced NF-κB activation in a reporter assay from 25-150 µM; reduces LPS-induced IL-6 production in RAW 264.7 cells; improves grip strength in the mdx mouse model of Duchenne muscular dystrophy at 30 mg/kg