Learn More
Cayman Chemical SR-17018, Purity: ≥98%, CAS: 2134602-45-0, 25 mg
SDP
Catalog No. 502936507
Encompass_Preferred
An analytical reference standard categorized as an opioid; increases latency to withdraw in the hot plate and warm water tail-flick assay without inducing respiratory depression in mice; induces place preference in a conditioned place preference test; induces antinociceptive tolerance after repeated administration in mice; intended for research and forensic applications
Catalog No. 50-293-6507 Supplier Cayman Chemical Supplier No. 2448025 MG
An analytical reference standard categorized as an opioid; increases latency to withdraw in the hot plate and warm water tail-flick assay without inducing respiratory depression in mice; induces place preference in a conditioned place preference test; induces antinociceptive tolerance after repeated administration in mice; intended for research and forensic applications