Learn More
Cayman Chemical ANTIBODY SR 17018 5mg
SDP
Supplier:  Cayman Chemical 244805 MG
Encompass_Preferred
An analytical reference standard categorized as an opioid; increases latency to withdraw in the hot plate and warm water tail-flick assay without inducing respiratory depression in mice; induces place preference in a conditioned place preference test; induces antinociceptive tolerance after repeated administration in mice; intended for research and forensic applications
Catalog No. 50-293-5504