Learn More
Cayman Chemical ANTIBODY SR 3335 5mg
SDP
Supplier:  Cayman Chemical 120725 MG
Encompass_Preferred
A selective inverse agonist of RORα, competitively inhibiting the binding of 25-hydroxycholesterol to the ligand binding domain (Ki = 220 nM) and inhibiting constitutive transactivation activity (IC50 = 480 nM); evokes RORα-dependent effects both in vitro and in vivo
Catalog No. 50-295-5806