Learn More
Cayman Chemical SR 4835, Purity: ≥98%, CAS: 2387704-62-1, 5 mg
SDP
Catalog No. 502916562
Encompass_Preferred
A Cdk12 and Cdk13 inhibitor; binds to Cdk12 and Cdk13 (Kds = 98 and 4.9 nM, respectively); inhibits Cdk12- and Cdk13-dependent phosphorylation of RNA polymerase II in MDA-MB-231 cells (EC50 = 105.5 nM); inhibits the proliferation of MDA-MB-231, MDA-MB-468, Hs 578T, and MDA-MB-436 breast cancer cells (EC50s = 15.5, 22.1, 19.9, and 24.9 nM, respectively); induces the DNA damage response and apoptosis in MDA-MB-231 from 30-90 nM; reduces tumor volume and decreases the number of mice with tumors greater than 1,000 mm3 in a PDX model of breast cancer at 20 mg/kg
Catalog No. 50-291-6562 Supplier Cayman Chemical Supplier No. 391585 MG
A Cdk12 and Cdk13 inhibitor; binds to Cdk12 and Cdk13 (Kds = 98 and 4.9 nM, respectively); inhibits Cdk12- and Cdk13-dependent phosphorylation of RNA polymerase II in MDA-MB-231 cells (EC50 = 105.5 nM); inhibits the proliferation of MDA-MB-231, MDA-MB-468, Hs 578T, and MDA-MB-436 breast cancer cells (EC50s = 15.5, 22.1, 19.9, and 24.9 nM, respectively); induces the DNA damage response and apoptosis in MDA-MB-231 from 30-90 nM; reduces tumor volume and decreases the number of mice with tumors greater than 1,000 mm3 in a PDX model of breast cancer at 20 mg/kg