Learn More
Boca Scientific Inc Antibody Stabilizer PBS Base, 125ml
Catalog No. NC0414486
Encompass
Antibody Stabilizer conserves the structure of proteins and antibodies to prevent loss of functionality due to storage. With refrigerated storage, Antibody Stabilizer prevents the need for freezing and thawing. Both TRIS-based and PBS-based Antibody Stabilizers are available, allowing the user to select the optimal buffer base for the assay.
Catalog No. NC0414486 Supplier Boca Scientific Inc Supplier No. 131 125
Antibody Stabilizer conserves the structure of proteins and antibodies to prevent loss of functionality due to storage. With refrigerated storage, Antibody Stabilizer prevents the need for freezing and thawing. Both TRIS-based and PBS-based Antibody Stabilizers are available, allowing the user to select the optimal buffer base for the assay.